Thermo Scientific Chemicals

D(+)-Glucurono-3,6-lactone, 99+%, Thermo Scientific Chemicals

Catalog number: 225920250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

D(+)-Glucurono-3,6-lactone, 99+%, Thermo Scientific Chemicals

Catalog number: 225920250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 225920250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32449-92-6
IUPAC Name
(2R)-2-[(2S,3R,4S)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde
Molecular Formula
C6H8O6
InChI Key
UYUXSRADSPPKRZ-SKNVOMKLSA-N
SMILES
O[C@@H](C=O)[C@H]1OC(=O)[C@@H](O)[C@H]1O
Specific optical rotation
+17.0° to +20.0° (20°C, 589 nm) (c=1, water)
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Melting point
170°C to 176°C
Titration excess NaOH
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text