D(+)-Glucurono-3,6-lactone, 99+%, Thermo Scientific Chemicals
D(+)-Glucurono-3,6-lactone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D(+)-Glucurono-3,6-lactone, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number 225925000
Price (USD)/ Each
232.65
Online exclusive
258.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
232.65
Online exclusive
258.00 
Save 25.35 (10%)
Add to cart
D(+)-Glucurono-3,6-lactone, 99+%, Thermo Scientific Chemicals
Catalog number225925000
Price (USD)/ Each
232.65
Online exclusive
258.00 
Save 25.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Name(2R)-2-[(2S,3R,4S)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde
Molecular FormulaC6H8O6
InChI KeyUYUXSRADSPPKRZ-SKNVOMKLSA-N
SMILESO[C@@H](C=O)[C@H]1OC(=O)[C@@H](O)[C@H]1O
Molecular Weight (g/mol)176.12
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+17.0° to +20.0° (20°C, 589 nm) (c=1, water)
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point170°C to 176°C
Titration excess NaOH>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only