Bacto Casitone is an animal origin (AO) pancreatic enzymatic digest of casein similar to Difco Casein Digest. While this supplementRead more
Have Questions?
Change view
Catalog Number
Quantity
225930
500 g
225910
10 kg
2 Options
Catalog number 225930
Price (USD)/ 500 g
193.00
-
Add to cart
Quantity:
500 g
Price (USD)/ 500 g
193.00
Add to cart
Bacto™ Casitone
Catalog number225930
Price (USD)/ 500 g
193.00
-
Add to cart
Bacto Casitone is an animal origin (AO) pancreatic enzymatic digest of casein similar to Difco Casein Digest. While this supplement provides a nutritional profile based on casein, the specific enzymatic digest performed for this product results in a particularly high content of amino acids and peptides of varying sizes, making it a more nutritious hydrolysate. Customers have successfully used this supplement for the production of Escherichia coli-based recombinant proteins.
Specifications
Certifications/ComplianceISO/DIS, 11014
FormPowder
Product TypeDehydrated Culture Media
Shelf Life1825 Days
FormatBottle
For Use With (Application)Production of Escherichia coli-based recombinant proteins
Product LineBacto™
Quantity500 g
SourceAnimal origin, casein
Unit Size500 g
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.