Maltotriose, 93%
Maltotriose, 93%
Maltotriose, 93%
Thermo Scientific Chemicals

Maltotriose, 93%

CAS: 1109-28-0 | C18H32O16 | 504.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 mg
Catalog number 225945000
Price (USD)
41.65
Special offer
Online exclusive
Ends: 26-Sep-2025
50.90
Save 9.25 (18%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 26-Sep-2025
50.90
Save 9.25 (18%)
Each
Add to cart
Chemical Identifiers
CAS1109-28-0
IUPAC Name2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC18H32O16
InChI KeyFYGDTMLNYKFZSV-UHFFFAOYNA-N
SMILESOCC1OC(OC2C(O)C(O)C(OC3C(O)C(O)C(O)OC3CO)OC2CO)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Specific optical rotation+155° to +165° (20°C, 589 nm) (c=1, water)
Infrared spectrumConforms
HPLC>=91 %
Water=<6 % (K.F.)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471069Certificate of AnalysisMay 17, 2025225940250, 225945000
A0456910Certificate of AnalysisOct 19, 2023225940250, 225945000
A0452198Certificate of AnalysisMay 02, 2023225940250, 225945000
A0417339Certificate of AnalysisDec 12, 2022225940250, 225945000
A0441256Certificate of AnalysisApr 29, 2022225940250, 225945000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection