Chemical Identifiers
CAS343336-76-5
IUPAC Name(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4S,5S)-3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyPVXPPJIGRGXGCY-VVLSOAGWNA-N
SMILESOC[C@H]1O[C@H](OC[C@H]2O[C@@](O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
View more