Thermo Scientific Chemicals

Xylitol, 99+%, Thermo Scientific Chemicals

Catalog number: 225980025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Xylitol, 99+%, Thermo Scientific Chemicals

Catalog number: 225980025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 225980025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
87-99-0
IUPAC Name
pentane-1,2,3,4,5-pentol
Molecular Formula
C5H12O5
InChI Key
HEBKCHPVOIAQTA-UHFFFAOYNA-N
SMILES
OCC(O)C(O)C(O)CO
Melting point
92°C to 96°C
Infrared spectrum
Conforms
GC
>=99.0 % (on dry substance)
Heavy metals (as Pb)
=<10 ppm
Arsenic (As)
=<3 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text