L(-)-Xylose, 99+%
Thermo Scientific Chemicals

L(-)-Xylose, 99+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 225990050
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.70 
Save 8.05 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.70 
Save 8.05 (14%)
Add to cart
L(-)-Xylose, 99+%
Catalog number225990050
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.70 
Save 8.05 (14%)
-
Add to cart
Chemical Identifiers
CAS609-06-3
IUPAC Name2,3,4,5-tetrahydroxypentanal
Molecular FormulaC5H10O5
InChI KeyPYMYPHUHKUWMLA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=99.0 %
Appearance (Color)White to off-white
Residue after ignition=<0.1 %
Specific optical rotationafter 24 h
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only