Opti-MEM™ Reduced Serum Medium, powder
Save up to 29% on your order of cell culture products. View promotion details >
Product Image
Gibco™

Opti-MEM™ Reduced Serum Medium, powder

Opti-MEM™ Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementationRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2260005010 x 1 L
2260013410 L
Catalog number 22600050
Price (USD)
371.00
Each
-
Add to cart
Quantity:
10 x 1 L
Customize this product
Price (USD)
371.00
Each
Add to cart

Opti-MEM™ Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change to growth rate or morphology. Opti-MEM™ is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Gibco™ Opti-MEM™ modifications for a range of cell culture applications.


This Opti-MEM™ is modified as follows:
With
• L-glutamine
• Phenol Red


The complete formulation is confidential. For more information, please contact Technical Services.

Gibco™ Opti-MEM™ is a unique media that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%.

Gibco™ Opti-MEM™ uses a sodium bicarbonate buffer system (2.4 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment and filtration at the time of preparation (see protocol for details).

Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.
Specifications
Cell LineSp2, AE-1, CHO, BHK-21, and HEK
Cell TypePrimary Fibroblasts
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco™, Opti-MEM™
Product TypeOpti-MEM I
Quantity10 x 1 L
Shelf Life30 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin
FormPowder
SterilitySterile-filtered
With AdditivesGlutamine, Phenol Red
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 30 months from date of manufacture
Please enter sku to load default questions

Frequently asked questions (FAQs)

Do you offer an alternative for Opti-MEM Reduced Serum Medium powder (Cat. No. 22600050)?

The direct alternative for Opti-MEM Reduced Serum Medium powder (Cat. No. 22600050) is Opti-MEM Reduced Serum Medium powder (Cat. No. 22600134). The product is the same but comes in a 10 L volume. Therefore, you might consider reconstituting the powder and then aliquoting the resulting media into smaller bottles.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How much Opti-MEM Reduced Serum Medium, powder (Cat. No. 22600050, 22600134) should I use to reconstitute the media?

We recommend reconstituting Opti-MEM Reduced Serum Medium, powder (Cat. No. 22600050, 22600134) at 13.592 g/L.

Is there any other difference between Opti-MEM I Reduced Serum Medium and Opti-MEM I Reduced Serum Medium, no phenol red besides the presence/absence of phenol red?

The formulation of Opti-MEM I Reduced Serum Medium and that of Opti-MEM I Reduced Serum Medium, no phenol red is the same except for the presence of phenol red in the former medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Where can I find the osmolality for my lot of Opti-MEM I Reduced Serum Medium?

We do provide osmolality information on the certificate of analysis. All lots of Opti-MEM I Reduced Serum Medium will meet the osmolality specification of ≥280 to ≤320 mOsm/kg.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I aliquot and freeze Opti-MEM I Reduced Serum Medium to conserve it longer? If so, for how long can it be stored?

Sorry, we don't have data to show that frozen Opti-MEM I Reduced Serum Medium has a longer shelf life. Actually, we recommend storing this medium at 2-8 degrees C in the dark. We do not recommend freezing the medium because it may precipitate.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link