L(+)-Norleucine, ≥99%, Thermo Scientific Chemicals
L(+)-Norleucine, ≥99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L(+)-Norleucine, ≥99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 226040010
Price (USD)/ Each
46.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.00 
Save 8.35 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
46.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.00 
Save 8.35 (15%)
Add to cart
L(+)-Norleucine, ≥99%, Thermo Scientific Chemicals
Catalog number226040010
Price (USD)/ Each
46.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.00 
Save 8.35 (15%)
-
Add to cart
Chemical Identifiers
CAS327-57-1
IUPAC Name(2S)-2-aminohexanoic acid
Molecular FormulaC6H13NO2
InChI KeyLRQKBLKVPFOOQJ-YFKPBYRVSA-N
SMILESCCCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=99.0 %
Other amino acids(TLC) Not detectable
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only