Thermo Scientific Chemicals

Stachyose hydrate, 97+%

Catalog number: 226085000
500 mg, Glass bottle, Each
Thermo Scientific Chemicals

Stachyose hydrate, 97+%

Catalog number: 226085000
500 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 226085000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
54261-98-2
IUPAC Name
(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-({[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-({[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}methyl)oxane-3,4,5-triol
Molecular Formula
C24H42O21
InChI Key
UQZIYBXSHAGNOE-CYDMKDQUNA-N
SMILES
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
HPLC
>=97.0 %
Water
=<12 % (K.F.)
Specific optical rotation
+129° to +140° (20°C, 589 nm) (c=4, water)
Solubility
(5 % in water) Clear to slightly hazy colorless
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text