Thermo Scientific Chemicals

Aristolochic acid, 96%, mixture of I and II, Thermo Scientific Chemicals

Catalog number: 226091000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

Aristolochic acid, 96%, mixture of I and II, Thermo Scientific Chemicals

Catalog number: 226091000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 226091000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
313-67-7
IUPAC Name
8-methoxy-6-nitro-2H-phenanthro[3,4-d][1,3]dioxole-5-carboxylic acid
Molecular Formula
C17H11NO7
InChI Key
BBFQZRXNYIEMAW-UHFFFAOYSA-N
SMILES
COC1=C2C=C(C3=C(C=C4OCOC4=C3C2=CC=C1)C(O)=O)[N+]([O-])=O
Appearance (Form)
Powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text