Dodecyl Sulfate, Sodium Salt 85%, Thermo Scientific Chemicals
Dodecyl Sulfate, Sodium Salt 85%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dodecyl Sulfate, Sodium Salt 85%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number 226145000
Price (USD)/ Each
61.65
Online exclusive
68.00 
Save 6.35 (9%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
61.65
Online exclusive
68.00 
Save 6.35 (9%)
Add to cart
Dodecyl Sulfate, Sodium Salt 85%, Thermo Scientific Chemicals
Catalog number226145000
Price (USD)/ Each
61.65
Online exclusive
68.00 
Save 6.35 (9%)
-
Add to cart
Chemical Identifiers
CAS151-21-3
IUPAC Namesodium dodecyl sulfate
Molecular FormulaC12H25NaO4S
InChI KeyDBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
GC>=80 %
Infrared spectrumConforms
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only