Thermo Scientific™

L-Photo-Methionine

Catalog number: 22615
Have Questions?
Thermo Scientific™

L-Photo-Methionine

Catalog number: 22615
Have Questions?
Catalog Number
22615
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
22615100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce L-Photo-Methionine is an analog of L-methionine that can be incorporated into proteins during synthesis and then light-activated to covalently crosslink interacting proteins in cells.

Features of L-Photo-Methionine:

In vivo labeling—incorporate photo-reactive group into proteins using normal cellular machinery of live cells
In vivo crosslinking—find interacting proteins in the native cellular environment
Increased specificity—crosslink interacting proteins correctly positioned at their interfaces within protein interaction domains
Efficient recovery—90% protein recovery in cell lysates after crosslinking
Compatible—crosslink proteins expressed in a wide variety of cell lines, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3 and C6
Easy to use—reagents are photo-stable under normal laboratory lighting, eliminating the need to work in the dark

Thermo Scientific L-Photo-Methionine is an analog of L-methionine that can be endogenously incorporated into the primary sequence of proteins during synthesis and then ultraviolet light (UV)-activated to covalently crosslink proteins within protein-protein interaction domains in their native environment within live cells. The powerful method enables both stable and transient protein interactions in cells to be studied and characterized without the use of completely foreign chemical crosslinkers and associated reagent solvents during the interaction experiment that can adversely aspects of the cell biology being studied.

When used in combination with specially formulated limiting media that is devoid of methionine, the photo-activatable derivative is treated like a naturally occurring amino acid by the protein synthesis machinery within the cell. As a result, it can be substituted for methionine in the primary sequence of proteins during catabolism and growth. Photo-methionine derivatives contain diazirine rings that activate when exposed to UV light to become reactive intermediates that form covalent bonds with nearby protein side chains and backbones. Naturally associating binding partners within the cell can be instantly trapped by photoactivation of the diazirine-containing proteins in the cultured cells. Crosslinked protein complexes can be detected by decreased mobility on SDS-PAGE followed by western blot detection, size exclusion chromatography, sucrose density gradient sedimentation or mass spectrometry.

Resources:

Photoreactive Amino Acid FAQ

Related Products
L-Photo-Leucine
Dulbecco's Modified Eagle's Limiting Medium (DMEM-LM)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Nonselective-NA (Metabolic labeling)
Cleavable
No
Description
L-Photo-Methionine
Molecular Weight (g/mol)
157.17
PEGylated
No
Spacer Arm Length
0.0 Å
Cell Permeability
Yes
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling, Metabolic Labeling
Crosslinker Type
Heterobifunctional
Reactive Moiety
Diazirine
Spacer
Short (<10 Å)
Form
Solid
Quantity
100 mg
Solubility
Water
Format
Standard
Water Soluble
Yes
Product Type
Crosslinker

Contents & Storage

Upon receipt store at 4°C protected from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text