Cytidine 5'-monophosphate hydrate, 99%
Cytidine 5'-monophosphate hydrate, 99%
Thermo Scientific Chemicals

Cytidine 5'-monophosphate hydrate, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 226210050
Price (USD)/ Each
250.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
295.00 
Save 44.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
250.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
295.00 
Save 44.35 (15%)
Add to cart
Cytidine 5'-monophosphate hydrate, 99%
Catalog number226210050
Price (USD)/ Each
250.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
295.00 
Save 44.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name{[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Molecular FormulaC9H14N3O8P
InChI KeyIERHLVCPSMICTF-NHBCCFCANA-N
SMILESNC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]1O
Molecular Weight (g/mol)323.20
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
UV spectrophotometry98 to 102 % (lambda max. 280 nm, pH 2)
Loss on drying=<7 % (105°C, 5 hrs)
Appearance (Form)Crystalline powder
HPLC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only