Inosine 5'-monophosphate, disodium salt hydrate, 97%
Inosine 5'-monophosphate, disodium salt hydrate, 97%
Thermo Scientific Chemicals

Inosine 5'-monophosphate, disodium salt hydrate, 97%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 226261000
Price (USD)/ Each
213.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
251.00 
Save 37.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
213.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
251.00 
Save 37.35 (15%)
Add to cart
Inosine 5'-monophosphate, disodium salt hydrate, 97%
Catalog number226261000
Price (USD)/ Each
213.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
251.00 
Save 37.35 (15%)
-
Add to cart
Chemical Identifiers
CAS352195-40-5
IUPAC Namedisodium [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-6,9-dihydro-1H-purin-9-yl)oxolan-2-yl]methyl phosphate
Molecular FormulaC10H11N4Na2O8P
InChI KeyAANLCWYVVNBGEE-WCYUCLFNNA-L
SMILES[Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1N=CNC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
UV spectrophotometrymolar absorptivity 12300 +/- 4%, on dry basis
Heavy metals (as Pb)=<20 ppm
UV spectrophotometry>=96.0 % (lambda max. 249 nm, pH 7)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only