Thermo Scientific Chemicals

Uridine 5'-monophosphate, disodium salt, hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 226290050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Uridine 5'-monophosphate, disodium salt, hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 226290050
5 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 226290050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3387-36-8
IUPAC Name
disodium [(2R,3S,4R,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Molecular Formula
C9H11N2Na2O9P
InChI Key
KURVIXMFFSNONZ-IJRYJGAHNA-L
SMILES
[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=CC(=O)NC1=O
Appearance (Color)
White
Infrared spectrum
Conforms
UV spectrophotometry
>=97.5 % (lambda max. 262 nm, pH 7)
Water
=<26 % (K.F.)
Ratio of Absorbances
0.71 to 0.77 (A250/A260)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text