Thermo Scientific Chemicals

2'-Deoxyguanosine 5'-monophosphate, sodium salt hydrate, 99%

Catalog number: 226342500
250 mg, Glass bottle, Each
Thermo Scientific Chemicals

2'-Deoxyguanosine 5'-monophosphate, sodium salt hydrate, 99%

Catalog number: 226342500
250 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 226342500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
52558-16-4
IUPAC Name
disodium [(2R,3S,5R)-5-(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)-3-hydroxyoxolan-2-yl]methyl phosphate
Molecular Formula
C10H12N5Na2O7P
InChI Key
CTPAMSRBXKGZCJ-WJQTVUBHNA-L
SMILES
[Na+].[Na+].NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](COP([O-])([O-])=O)O2)C(=O)N1
Appearance (Form)
Crystalline powder
Water
=<20 % (K.F.)
Infrared spectrum
Conforms
HPLC
>=98.5 %
Ratio of Absorbances
1.12 to 1.18 (A250/A260) c=1, water

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text