Thermo Scientific Chemicals

5-Bromouracil, 98%

Catalog number: 226440250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Bromouracil, 98%

Catalog number: 226440250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 226440250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
51-20-7
IUPAC Name
5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Formula
C4H3BrN2O2
InChI Key
LQLQRFGHAALLLE-UHFFFAOYSA-N
SMILES
BrC1=CNC(=O)NC1=O
Appearance (Form)
Crystalline powder
Appearance (Color)
Off-white to beige
HPLC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text