Kinetin, 99%, Thermo Scientific Chemicals
Kinetin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Kinetin, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 226500250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC NameN-[(furan-2-yl)methyl]-7H-purin-6-amine
Molecular FormulaC10H9N5O
InChI KeyQANMHLXAZMSUEX-UHFFFAOYSA-N
SMILESC(NC1=C2NC=NC2=NC=N1)C1=CC=CO1
Molecular Weight (g/mol)215.22
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO4>=98.5 %
Appearance (Color)White to beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only