This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
Atropine is a competitive and reversible antagonist of muscarinic acetylcholine receptors on both central and peripheral neurons
Atropine was originally isolated from the belladonna plant Atropa belladonna but is also found in other Solanaceae plants
Atropine is a drug that can be used to slow heart rate, decrease salivary secretions, and treat poisoning caused by cholinergic or muscarinic agents
Applications
It is used to block activation of muscarinic receptors in laboratory mice
It can be used in a laboratory setting to decrease oral and airway secretions in research animals
Atropine has been used to study heart rate and blood pressure variability in swine