Chemical Identifiers
CAS951-77-9
IUPAC Name4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Molecular FormulaC9H13N3O4
InChI KeyCKTSBUTUHBMZGZ-JORGKRSHNA-N
SMILESNC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1
View more