Thermo Scientific Chemicals

2'-Deoxyguanosine hydrate, 99+%

Catalog number: 226730050
5 g, Each
Thermo Scientific Chemicals

2'-Deoxyguanosine hydrate, 99+%

Catalog number: 226730050
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 226730050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207121-55-9
HPLC
>=99.0 %
UV spectrophotometry
molar absorptivity 13700 +/- 4%, on dry basis
Appearance (Form)
Crystalline powder
Water
=<8.0 % (K.F.)
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text