5-Aminouracil, 98%, Thermo Scientific Chemicals
5-Aminouracil, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Aminouracil, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
Catalog number 226820250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS932-52-5
IUPAC Name5-amino-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC4H5N3O2
InChI KeyBISHACNKZIBDFM-UHFFFAOYSA-N
SMILESNC1=CNC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Color)Beige to brown
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only