Thermo Scientific Chemicals

1-Naphthyl phosphate, monosodium salt monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 226890250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Naphthyl phosphate, monosodium salt monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 226890250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 226890250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81012-89-7
Infrared spectrum
Conforms
Solubility
(5 % in water) Clear colorless to light yellow
Appearance (Color)
White to off-white
Impurity
=<100 ppm Free 1-Naphthol
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text