Acetyl-β-methylcholine bromide, 99%, Thermo Scientific Chemicals
Acetyl-β-methylcholine bromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetyl-β-methylcholine bromide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 226970250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS333-31-3
IUPAC Name[2-(acetyloxy)propyl]trimethylazanium bromide
Molecular FormulaC8H18BrNO2
InChI KeyMMVPLEUBMWUYIB-UHFFFAOYNA-M
SMILES[Br-].CC(C[N+](C)(C)C)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Loss on drying=<1 %
Appearance (Color)White to off-white
Melting point149°C to 153°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only