Thermo Scientific Chemicals

1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals

Catalog number: 226995000
500 g, Each
Thermo Scientific Chemicals

1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals

Catalog number: 226995000
500 g, Each
Quantity
Catalog number: 226995000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6004-24-6
IUPAC Name
1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride
Molecular Formula
C21H40ClNO
InChI Key
NFCRBQADEGXVDL-UHFFFAOYSA-M
SMILES
O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1
Infrared spectrum
Conforms
Appearance (Form)
Powder
Water
4.5 to 5.5 %
Appearance (Color)
White
Titration
96.0 to 101.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text