Thermo Scientific™
UltiMate™ AFC-3000 Eluent Bottle
Catalog number: 2270.0026
Have Questions?
Thermo Scientific™
UltiMate™ AFC-3000 Eluent Bottle
Catalog number: 2270.0026
Have Questions?
Prepare and store up to 250mL eluent using the Thermo Scientific™ Dionex™ UltiMate™ AFC-3000 Eluent Bottle. For the Thermo Scientific™ Dionex™ UltiMate™ AFC-3000 Fraction Collector, the bottle can be used for either dispensing or collecting eluent for your analyses. The bottle includes screw cap for tubing guides. The UltiMate AFC-3000 Automated Fraction Collector is a rugged and flexible fraction collector, ideal for preparative reversed-phase or normal phase HPLC as well as purification of proteins, peptides, and nucleotides.
Catalog Number
2270.0026
Unit Size
Each
For Use With (Equipment)
Ultimate AFC-3000 Fraction Collector
Type
Eluent Bottle
Price (USD)
Full specifications
TypeEluent Bottle
Sample Capacity250 mL
For Use With (Equipment)Ultimate AFC-3000 Fraction Collector, Ultimate™ AFC-3000 Fraction Collector
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeFor Use With (Equipment)TypePrice (USD)
2270.0026Full specifications
EachUltimate AFC-3000 Fraction CollectorEluent BottleRequest A Quote
TypeEluent Bottle
Sample Capacity250 mL
For Use With (Equipment)Ultimate AFC-3000 Fraction Collector, Ultimate™ AFC-3000 Fraction Collector
Unit SizeEach
Showing 1 of 1
Prepare and store up to 250mL eluent using the Thermo Scientific™ Dionex™ UltiMate™ AFC-3000 Eluent Bottle. For the Thermo Scientific™ Dionex™ UltiMate™ AFC-3000 Fraction Collector, the bottle can be used for either dispensing or collecting eluent for your analyses. The bottle includes screw cap for tubing guides. The UltiMate AFC-3000 Automated Fraction Collector is a rugged and flexible fraction collector, ideal for preparative reversed-phase or normal phase HPLC as well as purification of proteins, peptides, and nucleotides.