Thermo Scientific Chemicals

Hexadecyltrimethylammonium bromide, 99+%, Thermo Scientific Chemicals

Catalog number: 227160025
2.5 kg, Plastic Drum, Each
Thermo Scientific Chemicals

Hexadecyltrimethylammonium bromide, 99+%, Thermo Scientific Chemicals

Catalog number: 227160025
2.5 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 227160025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-09-0
IUPAC Name
hexadecyltrimethylazanium bromide
Molecular Formula
C19H42BrN
InChI Key
LZZYPRNAOMGNLH-UHFFFAOYSA-M
SMILES
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
Solubility
(10% solution in water, after heating at 70°C)
Solubility
Clear to slightly hazy, colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text