Thermo Scientific Chemicals

D-Methionine, 99+%

Catalog number: 227210250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

D-Methionine, 99+%

Catalog number: 227210250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 227210250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
348-67-4
IUPAC Name
(2R)-2-amino-4-(methylsulfanyl)butanoic acid
Molecular Formula
C5H11NO2S
InChI Key
FFEARJCKVFRZRR-SCSAIBSYSA-N
SMILES
CSCC[C@@H](N)C(O)=O
Loss on drying
=<0.3 %
Sulfate (SO4)
=<300 ppm
Iron (Fe)
=<10 ppm
UV transmittance
>=98 % (c=3, H2O, 430 nm) 1 cm cell
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text