Thermo Scientific Chemicals

D(-)-Isoascorbic acid, 98%

Catalog number: 227275000
500 g, Each
Thermo Scientific Chemicals

D(-)-Isoascorbic acid, 98%

Catalog number: 227275000
500 g, Each
Quantity
Catalog number: 227275000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89-65-6
Appearance (Form)
Crystalline powder or crystals
Melting point
164°C to 172°C
Lead (Pb)
=<10 ppm
Appearance (Color)
White to light yellow
Loss on drying
=<0.4 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text