Methyl Viologen hydrate, 98%
Methyl Viologen hydrate, 98%
Methyl Viologen hydrate, 98%
Methyl Viologen hydrate, 98%
Thermo Scientific Chemicals

Methyl Viologen hydrate, 98%

CAS: 1910-42-5 | C12H14Cl2N2 | 257.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 227320050
Price (EUR)
230,35
Online Exclusive
271,00
Save 40,65 (15%)
Each
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
230,35
Online Exclusive
271,00
Save 40,65 (15%)
Each
Add to cart
Chemical Identifiers
CAS75365-73-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Appearance (Color)Beige to pink to brown
Titration Argentometric>=97.5 % (on dry substance)
Water=<18 % (K.F.)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0467142Certificate of AnalysisNov 06, 2024227320010, 227320050
A0461561Certificate of AnalysisApr 05, 2024227320010, 227320050
A0457002Certificate of AnalysisOct 19, 2023227320010, 227320050
A0417571Certificate of AnalysisDec 12, 2022227320010, 227320050
A0446898Certificate of AnalysisNov 08, 2022227320010, 227320050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 3
Acute dermal toxicity
Category 3
Acute Inhalation Toxicity - Dusts and Mists
Category 1
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Specific target organ toxicity - (repeated exposure)
Category 1
Acute aquatic toxicity
Category 1
Chronic aquatic toxicity
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H301 + H311 - Toxic if swallowed or in contact with skin

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H330 - Fatal if inhaled

H335 - May cause respiratory irritation

H372 - Causes damage to organs through prolonged or repeated exposure

H410 - Very toxic to aquatic life with long lasting effects

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Share catalog number, name or link

1x1 image pixel for data collection