Polyvinylpyrrolidone, average M.W. 1.300.000, K85-95, Thermo Scientific Chemicals
Polyvinylpyrrolidone, average M.W. 1.300.000, K85-95, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyvinylpyrrolidone, average M.W. 1.300.000, K85-95, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
500 g
1 kg
Packaging:
View all
Plastic Drum
Catalog number 227470025
Price (USD)/ Each
356.65
Online exclusive
396.00 
Save 39.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)/ Each
356.65
Online exclusive
396.00 
Save 39.35 (10%)
Add to cart
Polyvinylpyrrolidone, average M.W. 1.300.000, K85-95, Thermo Scientific Chemicals
Catalog number227470025
Price (USD)/ Each
356.65
Online exclusive
396.00 
Save 39.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9003-39-8
Molecular Formula(C6H9NO)n
SMILES*-CC(-*)N1CCCC1=O
MDL NumberMFCD01076626
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Water=<5 % (K.F.)
Sulfated ash=<0.1 %
Peroxides=<400 ppm (Titanyl sulphate method)
Hydrazine (N2H4)=<1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only