Thermo Scientific Chemicals

Polyvinylpyrrolidone, cross-linked, Thermo Scientific Chemicals

Catalog number: 227480250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Polyvinylpyrrolidone, cross-linked, Thermo Scientific Chemicals

Catalog number: 227480250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 227480250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
25249-54-1
Molecular Formula
(C6H9NO)n
SMILES
*-CC(-*)N1CCCC1=O
MDL Number
MFCD00149016
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Nitrogen (N)
11 to 12.8 % (as is)
Water
=<5 % (K.F.)
Peroxides
=<400 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text