1x1 image pixel for data collection
Polyvinylpyrrolidone, average M.W. 50.000, K30
Polyvinylpyrrolidone, average M.W. 50.000, K30
Polyvinylpyrrolidone, average M.W. 50.000, K30
Polyvinylpyrrolidone, average M.W. 50.000, K30
Thermo Scientific Chemicals

Polyvinylpyrrolidone, average M.W. 50.000, K30

CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 227541000
Price (USD)
52.65
Online Exclusive
58.20
Save 5.55 (10%)
Each
In stock
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
52.65
Online Exclusive
58.20
Save 5.55 (10%)
Each
Add to cart
Chemical Identifiers
CAS9003-39-8
Molecular Formula(C6H9NO)n
SMILES*-CC(-*)N1CCCC1=O
MDL NumberMFCD01076626
CAS9003-39-8
View more
SpecificationsSpecification SheetSpecification Sheet
K-value27 to 32 (1 % w/v in water at 25°C)
K-value27 to 32 (1 % w/v in water at 25°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection