Chemical Identifiers
CAS50-91-9
IUPAC Name5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H11FN2O5
InChI KeyODKNJVUHOIMIIZ-RRKCRQDMSA-N
SMILESOC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O
View more