Thermo Scientific Chemicals

N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride, 98%

Catalog number: 227740050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride, 98%

Catalog number: 227740050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 227740050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
911-77-3
IUPAC Name
5-[(diaminomethylidene)amino]-N-(4-nitrophenyl)-2-(phenylformamido)pentanamide hydrochloride
Molecular Formula
C19H23ClN6O4
InChI Key
DEOKFPFLXFNAON-UHFFFAOYNA-N
SMILES
Cl.NC(N)=NCCCC(NC(=O)C1=CC=CC=C1)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O
Appearance (Color)
White to yellow to light brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text