Thermo Scientific Chemicals

L-1-4'-Tosylamino-2-phenylethyl chloromethyl ketone, 99+%, Thermo Scientific Chemicals

Catalog number: 227800050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

L-1-4'-Tosylamino-2-phenylethyl chloromethyl ketone, 99+%, Thermo Scientific Chemicals

Catalog number: 227800050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 227800050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
402-71-1
IUPAC Name
N-[(2S)-4-chloro-3-oxo-1-phenylbutan-2-yl]-4-methylbenzene-1-sulfonamide
Molecular Formula
C17H18ClNO3S
InChI Key
MQUQNUAYKLCRME-INIZCTEOSA-N
SMILES
CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CC1=CC=CC=C1)C(=O)CCl
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
105°C to 109°C
Specific optical rotation
-86° to -88° (20°C, 589 nm) (c=1, abs. ethanol)
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text