Chemical Identifiers
CAS118-10-5
IUPAC Name(S)-[(2R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](quinolin-4-yl)methanol
Molecular FormulaC19H22N2O
InChI KeyKMPWYEUPVWOPIM-FRYPIZGFNA-N
SMILES[H][C@@]1(CN2CCC1C[C@]2([H])[C@@H](O)C1=CC=NC2=CC=CC=C12)C=C
View more