Thermo Scientific Chemicals

D(+)-Camphor, 97%, Thermo Scientific Chemicals

Catalog number: 227841000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

D(+)-Camphor, 97%, Thermo Scientific Chemicals

Catalog number: 227841000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 227841000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
464-49-3
IUPAC Name
(1R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Molecular Formula
C10H16O
InChI Key
DSSYKIVIOFKYAU-XVKPBYJWSA-N
SMILES
CC1(C)[C@H]2CC[C@@]1(C)C(=O)C2
Appearance (Color)
White
Specific optical rotation
+41° to +44° (20°C, 589 nm) (c=10, alcohol)
Appearance (Form)
Crystals
Melting point
174°C to 179°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text