Thermo Scientific Chemicals

Pararosaniline chloride, pure, high purity biological stain

Catalog number: 227885000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Pararosaniline chloride, pure, high purity biological stain

Catalog number: 227885000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 227885000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
569-61-9
IUPAC Name
hydrogen 4-[(4-aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
Molecular Formula
C19H18ClN3
InChI Key
JUQPZRLQQYSMEQ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC1=CC=C(C=C1)C(C1=CC=C(N)C=C1)=C1C=CC(=N)C=C1
Appearance (Form)
Crystalline powder
Dye content
>=85 %
Infrared spectrum
Conforms
Appearance (Color)
Green

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text