(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, 98%
(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, 98%
(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, 98%
(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, 98%
Thermo Scientific Chemicals

(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, 98%

CAS: 4272-74-6 | C14H22Cl2N2O3S | 369.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
50 mg
250 mg
Packaging:
Glass bottle
Catalog number 227950500
Price (USD)
-
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4272-74-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
Titration Argentometric>=97.5 %
Appearance of solution(5% soln. in methanol) clear, slightly colored
Specific optical rotation-9° to -10° (24°C, 589 nm) (c=5, H2O)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472197Certificate of AnalysisMay 17, 2025227952500, 227950010, 227950500
A0406024Certificate of AnalysisMar 11, 2024227952500, 227950010, 227950500
A0457398Certificate of AnalysisNov 03, 2023227952500, 227950010, 227950500
A0394194Certificate of AnalysisDec 15, 2022227952500, 227950010, 227950500
4 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection