ACES, 99%, extra pure
ACES, 99%, extra pure
Thermo Scientific Chemicals

ACES, 99%, extra pure

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
Catalog number 228000250
Price (USD)/ Each
46.65
Online exclusive
52.20 
Save 5.55 (11%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
46.65
Online exclusive
52.20 
Save 5.55 (11%)
Add to cart
ACES, 99%, extra pure
Catalog number228000250
Price (USD)/ Each
46.65
Online exclusive
52.20 
Save 5.55 (11%)
-
Add to cart
Chemical Identifiers
CAS7365-82-4
IUPAC Name2-[(carbamoylmethyl)amino]ethane-1-sulfonic acid
Molecular FormulaC4H10N2O4S
InChI KeyDBXNUXBLKRLWFA-UHFFFAOYSA-N
SMILESNC(=O)CNCCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
UV(5 % in water)
UVat 260 nm A: =<0.04
Titration with NaOH>=98.5 %
UVat 280 nm A: =<0.02
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only