Thermo Scientific Chemicals

ACES, 99%, extra pure

Catalog number: 228001000
100 g, Each
Thermo Scientific Chemicals

ACES, 99%, extra pure

Catalog number: 228001000
100 g, Each
Quantity
Catalog number: 228001000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7365-82-4
IUPAC Name
2-[(carbamoylmethyl)amino]ethane-1-sulfonic acid
Molecular Formula
C4H10N2O4S
InChI Key
DBXNUXBLKRLWFA-UHFFFAOYSA-N
SMILES
NC(=O)CNCCS(O)(=O)=O
Appearance (Color)
White
UV
(5 % in water)
UV
at 260 nm A: =<0.04
Titration with NaOH
>=98.5 %
UV
at 280 nm A: =<0.02

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text