ADA, 99%, for biochemistry
ADA, 99%, for biochemistry
Thermo Scientific Chemicals

ADA, 99%, for biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 228011000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS26239-55-4
IUPAC Name2-[(carbamoylmethyl)(carboxylatomethyl)azaniumyl]acetate
Molecular FormulaC6H9N2O5
InChI KeyQZTKDVCDBIDYMD-UHFFFAOYSA-M
SMILESNC(=O)C[NH+](CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<5 ppm
Sulfated ash=<0.1 %
Loss on drying=<0.5 % (110°C, 5 hrs)
UV(9 % in 1 M NaOH)
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only