Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure
This product ships via ground from US northeast.
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
250 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 228030010
Price (USD)/ Each
247.65
Online exclusive
275.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
247.65
Online exclusive
275.00 
Save 27.35 (10%)
Add to cart
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure
Catalog number228030010
Price (USD)/ Each
247.65
Online exclusive
275.00 
Save 27.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1185-53-1
IUPAC Namehydrogen 2-amino-2-(hydroxymethyl)propane-1,3-diol chloride
Molecular FormulaC4H12ClNO3
InChI KeyQKNYBSVHEMOAJP-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<5 ppm
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only