Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure

Catalog number: 228030051
5 kg, Plastic drum, Each
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure

Catalog number: 228030051
5 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 228030051
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1185-53-1
IUPAC Name
hydrogen 2-amino-2-(hydroxymethyl)propane-1,3-diol chloride
Molecular Formula
C4H12ClNO3
InChI Key
QKNYBSVHEMOAJP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC(CO)(CO)CO
Heavy metals (as Pb)
=<10 ppm
Iron (Fe)
=<5 ppm
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text