1x1 image pixel for data collection
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure
This product ships via ground from US northeast.
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure
Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure

CAS: 1185-53-1 | C4H11NO3·HCl | 157.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 228032500
Price (USD)
80.65
Special offer
Online exclusive
Ends: 26-Sep-2025
98.90
Save 18.25 (18%)
Each
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
80.65
Special offer
Online exclusive
Ends: 26-Sep-2025
98.90
Save 18.25 (18%)
Each
Add to cart
Chemical Identifiers
CAS1185-53-1
IUPAC Namehydrogen 2-amino-2-(hydroxymethyl)propane-1,3-diol chloride
Molecular FormulaC4H12ClNO3
InChI KeyQKNYBSVHEMOAJP-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<5 ppm
Infrared spectrumConforms
pH3.5 to 5.0 (1 % at 25°C)
Solubility(1 % in water) Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Share catalog number, name or link