Thermo Scientific Chemicals

Tranexamic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 228040500
50 g, Glass Bottle, Each
Thermo Scientific Chemicals

Tranexamic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 228040500
50 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 228040500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1197-18-8
IUPAC Name
4-(aminomethyl)cyclohexane-1-carboxylic acid
Molecular Formula
C8H15NO2
InChI Key
GYDJEQRTZSCIOI-UHFFFAOYSA-N
SMILES
NCC1CCC(CC1)C(O)=O
Infrared spectrum
Conforms
Loss on drying
=<1 %
pH
6.5 to 8.0 (5 % solution)
Appearance (Color)
White
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text