4-Aminobenzamidine dihydrochloride, 97%, Thermo Scientific Chemicals
4-Aminobenzamidine dihydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Aminobenzamidine dihydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 228050250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2498-50-2
IUPAC Namedihydrogen 4-aminobenzene-1-carboximidamide dichloride
Molecular FormulaC7H11Cl2N3
InChI KeyGHEHNICLPWTXJC-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Titration Argentometric>=96.0 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only