Pyridoxal 5-phosphate monohydrate, 99%
Pyridoxal 5-phosphate monohydrate, 99%
Thermo Scientific Chemicals

Pyridoxal 5-phosphate monohydrate, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 228170050
Price (USD)/ Each
158.65
Online exclusive
176.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
158.65
Online exclusive
176.00 
Save 17.35 (10%)
Add to cart
Pyridoxal 5-phosphate monohydrate, 99%
Catalog number228170050
Price (USD)/ Each
158.65
Online exclusive
176.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS41468-25-1
IUPAC Name[(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid
Molecular FormulaC8H10NO6P
InChI KeyNGVDGCNFYWLIFO-UHFFFAOYSA-N
SMILESCC1=NC=C(COP(O)(O)=O)C(C=O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow
Appearance (Form)Powder
HPLC>=98.5 %
Water=<10 %
Heavy metals (as Pb)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only